Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-MAP2K1 + MAP2K2 Rabbit Polyclonal Antibody

Supplier: BioVision

3518-100
10004-096EA 272.95 USD InStock
10004-096
Anti-MAP2K1 + MAP2K2 Rabbit Polyclonal Antibody
Antibodies
MAP kinase kinase 1 (Mek1), also known as mitogen activated protein kinase kinase, MAP kinase kinase, ERK kinase 1 or microtubule-associated protein 2 kinase, is a 43 kDa dual-specificity protein kinase that phosphorylates threonine and tyrosine residues on MAP kinases such as Erk 1 and 2. Mek1 and Mek2 are involved in a diverse array of cellular processes such as stress-activated response, apoptosis, cytokine-induced cell proliferation, and DNA recombination during meiosis. Mek1 and Mek2 are activated by phosphorylation on serine 218 and 222 by serine-threonine kinase, Raf-1, which is part of the p21ras signal transduction pathway.

Recommended Dilutions: Western Blot: 1:500-1000

Type: Primary
Antigen: MAP2K1 + MAP2K2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty