Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-TCF3/E2A Rabbit Polyclonal Antibody

Supplier: BioVision

3090-100
10003-346EA 252.35 USD InStock
10003-346
Anti-TCF3/E2A Rabbit Polyclonal Antibody
Antibodies
Heterodimers between TCF3 and tissue specific basic helix loop helix (bHLH) proteins play important roles in determining tissue specific cell fate during embryogenesis, like muscle or early B cell differentiation. Dimers bind DNA on E box motifs: 5' CANNTG 3'.Binds to the kappa E2 site in the kappa immunoglobulin gene enhancer.

Recommended Dilutions: Western Blot: 1 µg/ml

Type: Primary
Antigen: TCF3/E2A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty