Anti-KLF4 Rabbit Polyclonal Antibody

Supplier: BioVision
3089-200
10003-342EA 286.04 USD
10003-342
Anti-KLF4 Rabbit Polyclonal Antibody
KLF4 belongs to the erythroid Kruppel-like factor (EKLF) family that is involved in the differentiation of epithelial cells and may also function in skeletal and kidney development. KLF4 can act as both a repressor and activator of transcription. KLF4 regulates the expression of key transcription factors during embryonic development and is important in maintaining embryonic stem cells. It also contributes to the down-regulation of p53/TP53 transcription.

Recommended Dilutions: Western Blot: 1 µg/ml

Type: Primary
Antigen: KLF4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR