Anti-PRMT6 Rabbit Polyclonal Antibody

Supplier: BioVision
3086-100
10003-330EA 286.04 USD
10003-330
Anti-PRMT6 Rabbit Polyclonal Antibody
Protein arginine methylation is a form of post-translational modification that is catalyzed by protein arginine methyltransferases (PRMTs). PRMT6 catalyzes the sequential transfer of a methyl group from S-adenosyl-L-methionine to the side chain nitrogens of arginine residues within proteins to form methylated arginine derivatives and S-adenosyl-L-homocysteine. PRMT6 has been shown to act as a restriction factor for HIV replication.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: PRMT6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR