Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-MAPK1 Rabbit Polyclonal Antibody

Supplier: BioVision

3085R-100
10003-326EA 262.65 USD InStock
10003-326
Anti-MAPK1 Rabbit Polyclonal Antibody
Antibodies
The extracellular signal-regulated kinase 1 and 2 (Erk1 and Erk2) are closely related mitogen activated protein (MAP) kinases that are activated throμgh the extracellular stimulation by many growth factors, mitogens and differentiation promoting agents via a protein kinase cascade. Erk1/2 are activated approximately 1000 fold by phosphorylation of neighboring threonine and tyrosine residues by Mek1 and Mek2. Both sites must be phosphorylated for maximum activity. Erk1/2 kinases are ubiquitously distributed in the nervous system, as well as a broad range of cells and tissues. These two kinases represent proximal kinases in the classical MAP kinase cascade pathway which links growth and differentiation signals at the cell surface (throμgh tyrosine kinase) with transcription in the nucleus.

Recommended Dilutions: Western Blot: 1-4 µg/ml;

Type: Primary
Antigen: MAPK1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Sheep, Chicken

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty