Anti-CYP1A1 Rabbit Polyclonal Antibody

Supplier: BioVision
3084R-100
10005-506EA 289.52 USD
10005-506
Anti-CYP1A1 Rabbit Polyclonal Antibody
Cytochrome P450 is mainly found in liver, and can metabolize a wide range of compounds such as organic solvents, acetaminophen, dimethylnitrosoamine and aliphatic alcohol, which have certain toxicological effects in humans. Ethanol-induced cytochrome P450 IIE1 is considered to be one of the major causes of oxidative stress in the liver following ethanol treatment.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml) .However, the optimal conditions should be determined individually. The antibody detects ~58kDa cytochrome P450 IIE1 from samples of human, mouse, rat and dog origins.

Type: Primary
Antigen: CYP1A1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR