Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CALR Mouse Monoclonal Antibody [clone: S75]

Supplier: BioVision

3076-100
10003-318EA 303.85 USD InStock
10003-318
Anti-CALR Mouse Monoclonal Antibody [clone: S75]
Antibodies
Calreticulin is a calcium binding protein found in abundance in the endoplasmic reticulum and the sarcoplasmic reticulum. Like many other ER proteins, it has the conserved ER retention KDEL (Lys-Asp-Glu-Leu) sequence at its C-terminus. Calreticulin has also been detected in the nucleus and nuclear envelop. Recent studies sμggest that this soluble ER protein has a multifunctional role as it appears to be involved in calcium storage and regulation as well as having a molecular chaperone activity. Studies also sμggest its involvement in certain autoimmune diseases. Consistence with its multiple functions the calreticulin molecule appears to have a zonal character. The protein has both high and low affinity calcium binding sites.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: CALR
Clonality: Monoclonal
Clone: S75
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty