Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PPP3CA Rabbit Polyclonal Antibody

Supplier: BioVision

3075-100
10003-314EA 272.95 USD InStock
10003-314
Anti-PPP3CA Rabbit Polyclonal Antibody
Antibodies
Calcineurin is a calcium/calmodulin-dependent serine/threonine protein phosphatase. The active enzyme is a heterodimer consisting of a catalytic A subunit of 61 kDa and a myristylated regulatory B subunit of 19 kDa that binds calcium. The catalytic A subunit consists of an amino terminal catalytic domain, followed by calmodulin and B subunit binding sites, and a caboxyl-terminal autoinhibitory domain. Calnineurin is the target of immunosuppressant drμgs such as cyclosporin A and FK506. Calcineurin controls the translocation of nuclear factors of activated T cells (NFAT), a family of transcriptional activators that control the expression of cytokine genes essential to immune response, from the cytosol into the nucleus of activated T cells. Calcineurin has been proposed to play an important role in mediating calcium-activated cell death.

Recommended Dilutions: Western Blot: 1-4 µg/ml; for Immunoprecipitation: 10-20 µg/ml

Type: Primary
Antigen: PPP3CA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Hamster, Rabbit, Bovine

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty