Anti-SSEA4 Mouse Monoclonal Antibody [clone: 813-70]

Supplier: BioVision
3074-25
10003-312EA 251 USD
10003-312
Anti-SSEA4 Mouse Monoclonal Antibody [clone: 813-70]
This antibody reacts with the Stage-specific embryonic antigen-4 (SSEA-4) that is expressed upon the surface of human teratocarcinoma stem cells (EC), human embryonic germ (EG) cells) and human embryonic stem (ES) cells. No immunoreactivity is evident with undifferentiated murine EC, ES and EG cells. Expression of SSEA-4 is down-regulated following differentiation of human EC cells. In contrast, the differentiation of murine EC and ES cells may be accompanied by an increase in SSEA-4 expression.

Recommended Dilutions: 0

Type: Primary
Antigen: SSEA4
Clonality: Monoclonal
Clone: 813-70
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG3
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR