Anti-GZMB Mouse Monoclonal Antibody [clone: B18.1]

Supplier: BioVision
3073-100
10003-308EA 344.42 USD
10003-308
Anti-GZMB Mouse Monoclonal Antibody [clone: B18.1]
Granzyme B is a 27 kDa serine protease stored in granules of activated cytotoxic T cells and NK cells. Upon target cell contact, granzyme B is directionally exocytosed and assisted by perforin, enters the target cell. With its unique substrate specificity (granzyme B cleaves after Asp), granzyme B processes and activates various pro-caspases thereby inducing apoptosis in the target cell.

Recommended Dilutions: 0

Type: Primary
Antigen: Granzyme B
Clonality: Monoclonal
Clone: B18.1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR