Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CD40 Rabbit Polyclonal Antibody

Supplier: BioVision

3072-100
10003-304EA 272.95 USD InStock
10003-304
Anti-CD40 Rabbit Polyclonal Antibody
Antibodies
CD40 is a type I membrane protein found on the surface of B cells and primary carcinomas. It belongs to the TNF-R family. CD40 serves as the receptor for CD40 ligand (CD40L), a cytokine expressed on the cell surface of T cells. CD40 plays a critical role in B cell proliferation, antibody class switching, modulation of apoptosis in the germinal center throμgh interaction with T cells expressing CD40L, and activation of CD4+ T cells. In non-hemopoitic cells such as epidermal basal cells, CD40 ligation serves as the signal for inhibition of cell growth and induction of differentiation. The cytoplasmic domain of CD40 interacts with TRAF6, a TNFR associated factor homolog that mediates signaling from CD40. Ligation of CD40 with its ligand preferentially induces stress-activated protein kinases in B lymphocytes.

  • Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: CD40
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty