Anti-Fas/Apo1 Rabbit Polyclonal Antibody

Supplier: BioVision
3070R-100
10003-300EA 286.04 USD
10003-300
Anti-Fas/Apo1 Rabbit Polyclonal Antibody
Fas/Apo-1/CD95 is a cell surface type-I membrane glycoprotein with an apparent molecular weight of 44 kDa on SDS-PAGE. Fas is a member of the TNFR family that includes TNFR1, TNFR2, CD27, CD30, and CD40. Binding of Fas-ligand (FasL) to Fas or crosslinking of Fas by anti-Fas monoclonal antibodies leads to apoptosis in Fas expressing cells. Both Fas and TNF-R contain the 70 amino acids cytoplasmic “death domain” that is responsible for transmitting signal for apoptosis.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml; Immunohistochemistry: 5 µg/ml

Type: Primary
Antigen: Fas/Apo1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR