Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-SSEA3 Rat Monoclonal Antibody [clone: MC-631]

Supplier: BioVision

3065-25
10003-298EA 211.15 USD InStock
10003-298
Anti-SSEA3 Rat Monoclonal Antibody [clone: MC-631]
Antibodies
This antibody reacts with the Stage-specific embryonic antigen-3 (SSEA-3). The epitope is expressed on the surface of human teratocarcinoma stem cells , embryonic carcinoma (EC) cells, human embryonic germ(EG) cells and human embryonic stem (ES) cells. Expression of SSEA-3 diminishes with differentiation of human EC cells.

Recommended Dilutions: Flow (cell Surface; IHC, IP, Western Blot

Type: Primary
Antigen: SSEA3
Clonality: Monoclonal
Clone: MC-631
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgM
Reactivity: Human, Mouse

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty