Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CD15 Mouse Monoclonal Antibody [clone: MC-480]

Supplier: BioVision

3063-25
10003-296EA 211.15 USD InStock
10003-296
Anti-CD15 Mouse Monoclonal Antibody [clone: MC-480]
Antibodies
Stage-Specific Embyronic Antigen-1(SSEA-1), also referred to as Lewis X (LeX), is expressed on murine and human germ cells, murine embryonic stem (ES) cells and murine embryonal carcinoma (EC) cells . Expression levels differ between mouse and human with decreasing expression upon differentiation in mice and increased expression upon differentiation in humans.

Recommended Dilutions: Flow (cell Surface; IF/ICC, IHC, IP

Type: Primary
Antigen: CD15
Clonality: Monoclonal
Clone: MC-480
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty