Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-TNFRSF10B Rabbit Polyclonal Antibody

Supplier: BioVision

3062-100
10003-292EA 252.35 USD InStock
10003-292
Anti-TNFRSF10B Rabbit Polyclonal Antibody
Antibodies
DR5 is the DD containing receptor for TRAIL, Apo2, TRAIL-R2, TRICK2, or KILLER. DR5 transcript is widely expressed in normal tissues and in many types of tumor cells. Like DR3 and TNFR1, DR5 mediate apoptosis by FADD dependent pathway. Over-expression of DR5 induces apoptosis and activates NFkB.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: TNFRSF10B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty