Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CFLAR Rabbit Polyclonal Antibody

Supplier: BioVision

3058-100
10003-278EA 252.35 USD InStock
10003-278
Anti-CFLAR Rabbit Polyclonal Antibody
Antibodies
c-FLIP was cloned by several labs independently and termed Casper, I-FLICE, FLAME-1, CASH AND CLARP. c-FLIP contains two death effector domains and has sequence homology to caspase-8. c-FLIP interacts with adapter protein FADD and caspaser-8 and -10, and potently inhibits apoptosis induced by all known death receptors. Two splice variants of c-FLIP have been identified and termed FLIPl and FLIPs

Recommended Dilutions: Western Blot: 0.5-4 µg/ml; immunofluorescence

Type: Primary
Antigen: CFLAR
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype: IgG
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty