Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-DR3 Rabbit Polyclonal Antibody

Supplier: BioVision

3057R-100
10003-274EA 252.35 USD InStock
10003-274
Anti-DR3 Rabbit Polyclonal Antibody
Antibodies
Apoptosis can be induced by certain cytokines including TNF and Fas in the TNF superfamily throμgh interaction with their death domain (DD) containing receptors, TNFR1 and Fas. A member of the TNF receptor family was recently identified and designated DR3 (also called Wal-1, Apo3, TRAMP and LARD). The ligand for this novel death receptor has been defined as TWEAK (also termed Apo3L). Like TNFR1, overexpression of DR3 triggers apoptosis and NF-kB activation. Activation of DR3 results in recruitment of TRADD (a DD containing adapter molecule). TRADD associates with FADD, TRAF2, and RIP to activate the signaling complex for NF-kB activation. DR3 activates apoptosis throμgh TRADD, FADD, and caspase-8. DR3 is highly expressed in the tissues enriched in lymphocytes including peripheral blood leucocytes (PBL), thymus and spleen.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml; Immunohistochemistry: 5 µg/ml

Type: Primary
Antigen: DR3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty