Anti-MBD4 Rabbit Polyclonal Antibody

Supplier: BioVision
3056-100
10003-270EA 286.04 USD
10003-270
Anti-MBD4 Rabbit Polyclonal Antibody
MBD4 (methyl-CpG-binding domain protein 4) belongs to the MBD family of nuclear proteins that bind to methylated CpG sites and represses gene transcription. Human proteins MECP2, MBD1, MBD2, and MBD4, except MBD3 bind specifically to methylated DNA. MBD4 exhibits thymine glycosylase activity and is known to play an important function in DNA mismatch repair and apoptosis.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: MBD4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR