Anti-TNFA Rabbit Polyclonal Antibody

Supplier: BioVision
3053R-100
10003-266EA 286.04 USD
10003-266
Anti-TNFA Rabbit Polyclonal Antibody
Tumor Necrosis Factor-alpha (TNF-α) is a potent lymphoid factor that exerts cytotoxic effects on a wide range of tumor cells and certain other target cells. Murine TNF-α is a 17.5 kDa protein containing 156 amino acid residues.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: TNFA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR