Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-FOXO4 Rabbit Polyclonal Antibody

Supplier: BioVision

3270-100
10003-714EA 334.75 USD InStock
10003-714
Anti-FOXO4 Rabbit Polyclonal Antibody
Antibodies
The forkhead family of transcription factors is involved in tumorigenesis in rhabdomyosarcoma and acute leukemias. Within the family, three members (FKHR, AFX, AND FKHRL1) have sequence similarity to the nematode homolog Daf 16, recently shown to mediate signaling via a pathway involving IGFR1, P13-K and Akt. There are three potential Akt phosphorylation sites on the FKHR proteins. One site, FKHR Ser256 is found in the Forkhead domain and the other two are outside this domain. Recent studies show that phosphorylation of FKHR family members by Akt promotes cell survival. Phosphorylation regulates FKHR nuclear translocation and target gene transcription. Insulin also stimulates FKHR phosphorylation at Ser253.

Recommended Dilutions: Western Blot: 1-2 µg/ml

Type: Primary
Antigen: Phospho-FoxO4/AFX
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty