Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-BAD Rabbit Polyclonal Antibody

Supplier: BioVision

3269-100
10003-712EA 334.75 USD InStock
10003-712
Anti-BAD Rabbit Polyclonal Antibody
Antibodies
Bad is a member of the Bcl-2 family protein that selectively dimerize with Bcl-xL and Bcl-2 resulting in cell death. Survival factors such as IL-3 can inhibit the apoptotic activity of Bad by activating intracellular signaling pathways that result in the phosphorylation of Bad at Ser112 and Ser 136. Phosphorylation at these sites results in the binding of Bad to 14-3-3 proteins and the inhibition of Bad binding to Bcl-xL and Bcl-2. Recently, Akt has also been shown to promote cell survival via its ability to phosphorylate Bad at Ser136.

Recommended Dilutions: Western Blot: 1-2 µg/ml; Immunoprecipitation: 5-20 µg/ml

Type: Primary
Antigen: BAD
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty