Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-FATP4 Rabbit Polyclonal Antibody

Supplier: BioVision

3268-100
10003-708EA 252.35 USD InStock
10003-708
Anti-FATP4 Rabbit Polyclonal Antibody
Antibodies
FATP4 is a transmembrane protein that is involved in the uptake and metabolism of long chain fatty acids. FATP4 has acyl-CoA synthase activity and contains an AMP-binding motif. FATP4 is linked to insulin resistance and metabolic disorders.

Recommended Dilutions: Western Blot: 1-4 µg/ml

Type: Primary
Antigen: FATP4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty