Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-LMNA Rabbit Polyclonal Antibody

Supplier: BioVision

3267-100
10003-704EA 252.35 USD InStock
10003-704
Anti-LMNA Rabbit Polyclonal Antibody
Antibodies
Lamins are nuclear membrane structural components that are important in maintaining normal cell functions such as cell cycle control, DNA replication and chromatin organization. Lamin A is specifically cleaved by caspase-6 and therefore serves as a marker for caspase-6 activation. During apoptosis the 70 kDa lamin A is cleaved to a large (40-45 kDa) and small (23 kDa) fragment. The cleavage of lamins results in nuclear disregulation and cell death.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml; Immunohistochemistry: 15-20 µg/ml

Type: Primary
Antigen: LMNA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty