Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-TCTN2 Rabbit Polyclonal Antibody

Supplier: BioVision

3262-100
10003-694EA 252.35 USD InStock
10003-694
Anti-TCTN2 Rabbit Polyclonal Antibody
Antibodies
TCTN2 is a member of the Tectonic protein family. Tectonic proteins are transmembrane proteins that regulate the Hedgehog (Hh)-signaling pathway. Defects in TCTN2 can lead to Meckel syndrome type 8. This disorder is characterized by a combination of renal cysts and variable associated features which includes developmental anomalies of the central nervous system, hepatic ductal dysplasia and cysts, and polydactyly.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: TCTN2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty