Anti-DLGAP1 Rabbit Polyclonal Antibody

Supplier: BioVision
3261-100
10003-692EA 332.74 USD
10003-692
Anti-DLGAP1 Rabbit Polyclonal Antibody
Death Associated Protein 1 (DAP1) is a 15 kDa protein that functions as a positive mediator of cell death initiated by interferon-gamma. The DAP1 protein is praline-rich and possesses one SH3 binding motif, as well as several consensus protein kinase phosphorylation sites. The protein is localized in the cytoplasma, however the detailed mechanism of its proapoptotic function is unclear.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: DLGAP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR