Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CASP8 Mouse Monoclonal Antibody [clone: C502S]

Supplier: BioVision

3258-100
10005-276EA 375.95 USD InStock
10005-276
Anti-CASP8 Mouse Monoclonal Antibody [clone: C502S]
Antibodies
Caspase-8, a member of the caspase-family of proteases, plays a key role in mediating Fas (CD95) and TNF induced apoptosis. Caspase-8 is synthesized as inactive pro-enzyme and activation of the enzyme involves proteolytic cleavage that leads to the release of the active p18 and p10 subunits. Activated caspase-8 is able to cleave and activate downstream caspases, such as caspase-3, -6, -7 and a death agonist member of the Bcl-2/Bcl-xL family, Bid, leading to apoptosis.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml; Immunoprecipitation: 10 µg/ml

Type: Primary
Antigen: CASP8
Clonality: Monoclonal
Clone: C502S
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Mouse
Isotype: IgG1
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty