Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-AKT Rabbit Polyclonal Antibody

Supplier: BioVision

3257-100
10005-274EA 350.2 USD InStock
10005-274
Anti-AKT Rabbit Polyclonal Antibody
Antibodies
Akt, also referred to as PKB and RAC, is the cellular homologue of the viral oncogene v-Akt. This protein kinase is activated by insulin and various growth factors and functions in a pathway involving PI3 kinase as activation is sensitive to wortmanin. Recent evidence sμggests that Akt functions to promote cell survival by actively inhibiting apoptosis, althoμgh the pathways involved have not been identified. Akt is activated by phospholipid binding and phosphorylation within the activation loop at Thr308 and within the C-terminus at Ser473.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml; Immunoprecipitation: 10-20 µg/ml

Type: Primary
Antigen: AKT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty