Anti-CASP6 Rabbit Polyclonal Antibody

Supplier: BioVision
3256-100
10005-270EA 286.04 USD
10005-270
Anti-CASP6 Rabbit Polyclonal Antibody
Caspase-6 is one of the major executioner caspases functioning in cellular apoptotic processes. Similar as other caspases, caspase-6 is also synthesized as an inactive pro-enzyme that is processed in cells undergoing apoptosis. Caspase-6 has also been shown involving in the proteolysis of poly (ADP-ribose) polymerase (PARP) and nuclear lamin A.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml; and Immunohistochemistry: 5 µg/ml

Type: Primary
Antigen: CASP6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR