Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-HSD17B10 Rabbit Polyclonal Antibody

Supplier: BioVision

3255-100
10005-578EA 252.35 USD InStock
10005-578
Anti-HSD17B10 Rabbit Polyclonal Antibody
Antibodies
ERAB/HCD2 (Hydroxyacyl-Coenzyme A dehydrogenase) is a mitochondrial protein that catalyzes the oxidation of straight-chain 3-hydroxyacyl-CoAs as part of the beta-oxidation pathway. ERAB is characterized as a NAD+-dependent dehydrogenase that overexpressed in neurons affected with Alzheimer’s disease.

Recommended Dilutions: Western Blot: 1-4 µg/ml

Type: Primary
Antigen: HSD17B10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty