Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IkBalpha Rabbit Polyclonal Antibody

Supplier: BioVision

3252-100
10003-686EA 252.35 USD InStock
10003-686
Anti-IkBalpha Rabbit Polyclonal Antibody
Antibodies
IkB-alpha protein is a NF-kB inhibitory protein. It binds NF-kB and retains the complex in the cytoplasm, thereby preventing it from entering into nucleus and functioning as a transcription factor. Upon receiving a variety of signals, IkB-alpha undergoes phosphorylation at serine residues by an ubiquitin-dependent protein kinases and finally degraded by the proteasome. Free NF-kB then binds DNA and affects gene expression.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: IkBalpha
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty