Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-TLR4 Rabbit Polyclonal Antibody

Supplier: BioVision

3251-100
10003-684EA 272.95 USD InStock
10003-684
Anti-TLR4 Rabbit Polyclonal Antibody
Antibodies
TLR4 (Toll-like receptor 4) is a mammalain homologue of the Drosophila toll protein. The human TLR4 is a type-1 transmembrane protein containing 799 amino acid residues. The extracellular domain has leucine-rich repeats and the intracellular domain is similar to that of IL-1 receptor. TLR4 is a receptor for lipopolysaccharide (LPS). In mice that are genetically tolerant to endotoxin, mutations have been identified in the intracellular domain of TLR4 that abolish LPS responsiveness. TLR4 uses a mechanism similar to that of the IL-1 receptor for signal transduction that leads to activation of NF-kB.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml;The antibody is also suitable for flow cytometric analysis of cell surface expression of TLR4

Type: Primary
Antigen: TLR4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty