Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-GAI2 Rabbit Polyclonal Antibody

Supplier: BioVision

3249-100
10003-678EA 252.35 USD InStock
10003-678
Anti-GAI2 Rabbit Polyclonal Antibody
Antibodies
Gαi2 is a 40-41 kDa alpha-subunit of heterotrimeric protein that shares sequence homology with Gαi3 and Gαi1. All these G proteins can be ADP-ribosylated at the carboxy terminal Cys of the CAAX sequence, interfering with receptor coupling. Gαi2 is expressed widely in tissues and cell lines. Activation of PLC-β by Gαi-coupled receptors is the result of release of Gβg subunits from Gαi. Gαi2 regulates potassium channel activation. Mutation at Arg179 has been identified in tumors. This and the Q205L mutant inhibit adenylyl cyclase.

Recommended Dilutions: Western Blot: at 0.5-4 µg/ml; immunoprecipitation

Type: Primary
Antigen: Gai2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty