Anti-EZH2 Rabbit Polyclonal Antibody

Supplier: BioVision
3242-100
10003-654EA 286.04 USD
10003-654
Anti-EZH2 Rabbit Polyclonal Antibody
Enhancer of zeste homolog 2 (Ezh2) is a member of the Polycomb group (PcG). The PcG proteins are involved in maintaining the silenced state of several developmentally regulated genes and contribute to the maintenance of cell identity, cell cycle regulation, and oncogenesis. Enhancer of zeste homolog 2 (Ezh2) contains four conserved regions including domain I, domain II, and a cysteine-rich amino acid stretch that precedes the carboxy-terminal SET domain. Ezh2 also plays a role as primary effector and as a mediator of tumorigenesis.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: EZH2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR