Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-MAP3K5 Rabbit Polyclonal Antibody

Supplier: BioVision

3128-100
10003-410EA 272.95 USD InStock
10003-410
Anti-MAP3K5 Rabbit Polyclonal Antibody
Antibodies
ASK1 (Apoptosis Signal-regulating Kinase 1) activates two different subgroups of MAPKK, MKK4 and MKK6, which in turn activate c-Jun N-terminal Kinase (JNK) and p38 MAP Kinase. ASK1 is activated by TNFR and Fas throμgh the interaction with members of the TRAF Family and the Fas-associated protein Daxx. Overexpression of ASK1 induces apoptosis. However, a catalytically inactive form of ASK1 has been shown to inhibit TNF-α induced apoptosis.

Recommended Dilutions: Western Blot: 1-4 µg/ml

Type: Primary
Antigen: MAP3K5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty