Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-TNFR1 Rabbit Polyclonal Antibody

Supplier: BioVision

3125-100
10003-406EA 288.4 USD InStock
10003-406
Anti-TNFR1 Rabbit Polyclonal Antibody
Antibodies
Tumor necrosis factor receptor 1 and 2 (TNF-R1 and TNF-R2) are 55 and 75 kDa. While TNF-R1 and TNF-R2 share 28% sequence homology in the extracellular domains, their intracellular domains lack sequence homology, sμggesting that they differ in their internal signal transduction pathways. TNF-R1 contains an approximately 80 amino acid death domain near its carboxy terminus capable of transmitting an apoptotic signal throμgh its interaction with TRADD (TNF-R1 associated death domain protein), and subsequent interactions with FADD. TNF-R1 can also activate the transcription factor NFkB via TRAF2 (TNF receptor associated factor 2). The cytoplasmic domain of TNF-R1 can directly interact with Jak kinase, thereby activating the JAK/STAT signal transduction cascade.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml; in Immunohistochemistry: 10-20 µg/ml

Type: Primary
Antigen: TNFR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Hamster, Bovine, Rabbit, Monkey

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty