Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-BCL2L11 Rabbit Polyclonal Antibody

Supplier: BioVision

3124-100
10003-402EA 252.35 USD InStock
10003-402
Anti-BCL2L11 Rabbit Polyclonal Antibody
Antibodies
Members of the Bcl-2 family proteins all contain Bcl-2 homology domains including BH1, BH2, BH3, and BH4. The BH3 domain is known to be important for the cell killing activity of Bcl-2 members including Bad, Bax, Bid, Bik, and the recently identified BH3 domain containing protein designated Bim/BOD. Bim/BOD interacts with diverse members of the Bcl-2 sub-family including Bcl-2, Bcl-x, and Bcl-w, to induce apoptosis. There are three splice variants: BimL, BimS, and BimEL, all of which share homology in their C-terminal BH3 domains. Of the isoforms, BimS is the most potent inducer of apoptosis. The mRNA of Bim is ubiquitously expressed in multiple tissues and cell lines.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: BCL2L11
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty