Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-DFFB Rabbit Polyclonal Antibody

Supplier: BioVision

3049-100
10003-262EA 289.57 USD InStock
10003-262
Anti-DFFB Rabbit Polyclonal Antibody
Antibodies
DFF is composed of two subunits: DFF40/CAD and DFF45/ICAD. Human DFF45 and its mouse homologue ICAD are the inhibitors of DFF40 and CAD, respectively. Cleavage of DFF45, which is mediated by caspase-3, leads to DFF40’s activation as a nuclease. Activation of DFF40/CAD can lead to DNA fragmentation, a hallmark of apoptosis. DFF40 mRNA is expressed in limited number of human tissues: pancreas, spleen, prostate, and ovary.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml; Immunohistochemistry: 2-8 µg/ml

Type: Primary
Antigen: DFFB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty