Anti-DFFA Rabbit Polyclonal Antibody

Supplier: BioVision
3048-100
10003-260EA 309.39 USD
10003-260
Anti-DFFA Rabbit Polyclonal Antibody
Apoptosis occurs during normal cellular development and involves dramatic changes in cellular structure. Disruption of apoptosis may contribute to cancer as well as many other types of diseases. Recently, a human DNA fragmentation factor (DFF45) was identified. DFF is composed of two subunts: DFF40 and DFF45. Cleavage of DFF45, which is mediated by caspase-3, leads to DFF40’s activation as a nuclease. Activation of DFF40 can lead to DNA fragmentation, a hallmark of apoptosis.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: DFFA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR