Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-TNF-related Apoptosis-inducing Ligand Rabbit Polyclonal Antibody

Supplier: BioVision

3045-100
10003-258EA 252.35 USD InStock
10003-258
Anti-TNF-related Apoptosis-inducing Ligand Rabbit Polyclonal Antibody
Antibodies
TRAIL (TNF-related apoptosis-inducing ligand), a member of the TNF ligand family, is a type II membrane protein and expressed in a variety of human tissues. Two death domains containing receptors DR4 and DR5 have been identified as the receptors for TRAIL and two decoy receptors designated DcR1 and DcR2 inhibit TRAIL function. The multiple receptors sμggest a complex role for regulation of signaling by the cytokine. TRAIL induces apoptosis and NF-kB activation in many tissues and cells.

Recommended Dilutions: Western Blot: 0.5-1 µg/ml; ELISA: 2-3 µg/ml; Neutralization: 5-10 µg/ml

Type: Primary
Antigen: TNF-related Apoptosis-inducing Ligand
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty