Anti-BZRAP1 Rabbit Polyclonal Antibody

Supplier: BioVision
AB2305-100
10005-496EA 393.47 USD
10005-496
Anti-BZRAP1 Rabbit Polyclonal Antibody
Antibodies
PBR (Peripheral-type Benzodiazepine Receptor) is a ubiquitous protein (18 kDa) found in most steroidogenic tissues. It is located primarily in the outer mitochondrial membrane of steroid-producing tissues and is found in association with a 34 kDa voltage-dependent anion channel protein (VDAC). PBR is involved in the regulation of cholesterol transport from the outer to the inner mitochondrial membrane. It is thoμght to be part of the mitochondrial permeability transition pore (PT) and to be involved in apoptosis.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml; Immunohistochemistry: 10-20 µg/ml

Type: Primary
Antigen: BZRAP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR