Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-BOK Rabbit Polyclonal Antibody

Supplier: BioVision

3043-100
10005-494EA 293.55 USD InStock
10005-494
Anti-BOK Rabbit Polyclonal Antibody
Antibodies
Bok (Bcl-2-related ovarian killer) is a pro-apoptotic Bcl-2 family protein identified in the ovary based on its dimerization with the anti-apoptotic protein Mcl-1. In addition to the Bcl-2 homology (BH) domains 1 and 2 and the transmembrane sequence, Bok also has a BH3 domain believed to be important for dimerization with selective anti-apoptotic Bcl-2 proteins and cell killing. Bok interacts strongly with some (Mcl-1, BHRF1, and Bfl-1) but not other (Bcl-2, Bcl-xL, and Bcl-w) anti-apoptotic members. In addition, cell killing induced by Bok was suppressed following coexpression with Mcl-1 and BHRF1, but not with Bcl-2, further indicating that Bok heterodimerized only with selective anti-apoptotic Bcl-2 proteins. Bok was highly expressed in the ovary, testis and uterus.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml;

Type: Primary
Antigen: BOK
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty