Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-FADD Rabbit Polyclonal Antibody

Supplier: BioVision

3039-100
10005-490EA 283.25 USD InStock
10005-490
Anti-FADD Rabbit Polyclonal Antibody
Antibodies
Fas-associating death domain containing protein (FADD), also known as mediator of receptor induced toxicity (MORT1), is a 23 kDa cytosolic protein with an apparent molecular weight of 28 kDa on SDS-PAGE. The protein is involved in both Fas and TNF-R induced cell death pathway. FADD specifically binds to Fas to initiate the apoptotic signalling cascade. Overexpression of FADD induces apoptosis.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml; can also be used in immunoprecipitation

Type: Primary
Antigen: FADD
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Monkey

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty