Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-P53 Rabbit Polyclonal Antibody

Supplier: BioVision

3036R-100
10005-480EA 252.35 USD InStock
10005-480
Anti-P53 Rabbit Polyclonal Antibody
Antibodies
p53, a 53 kDa protein, binds to a DNA consensus sequence, the p53 response element, and regulates normal cell cycle events by activating transcription of genes involved either in progression throμgh the cell cycle, or causing arrest in G1 when the genome is damaged. In most transformed and tumor cells the concentration of p53 is increased 5-1000 fold over the concentration in normal cells, principally due to the increased half-life (4 hrs) compared to that of wild-type (20 min). p53 localizes in the nucleus, but is detectable at the plasma membrane during mitosis and certain mutations also modulate cytoplasmic/nuclear distribution. p53 downregulates Bcl-2 expression and upregulates Bax expression, but may not always be necessary for apoptosis.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: P53
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Dog, Rabbit

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty