Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-BCL2 Rabbit Polyclonal Antibody

Supplier: BioVision

3033-100
10005-470EA 283.25 USD InStock
10005-470
Anti-BCL2 Rabbit Polyclonal Antibody
Antibodies
Bcl-2 and its homologs belong to a family of proteins that either promotes or prevent apoptosis. Bcl-2 alpha and beta are the alternatively spliced isoforms of 25 kDa and 22 kDa mitochondrial integral membrane proteins that inhibit apoptosis.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml; Immunoprecipation: 4-8 µg/ml; and Immunohistochemistry: 10-20 µg/ml

Type: Primary
Antigen: BCL2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty