Anti-BAX Rabbit Polyclonal Antibody

Supplier: BioVision
3032-100
10003-256EA 286.04 USD
10003-256
Anti-BAX Rabbit Polyclonal Antibody
The Bcl-2 (B-cell leukemia/lymphoma 2) family proteins function to either inhibit (Bcl-2 and Bcl-XL) or promote (Bax and Bak) apoptosis. Bax contains the conserved Bcl-2 homology 1 (BH1) and 2 (BH2) domains that allow for its homodimerization or heterodimerization with Bcl-2. When Bcl-2 is in excess, apoptosis is inhibited. However, if Bax levels increase in response to a death signal, the cell is pushed toward death.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml; Immunoprecipitation: 10-20 µg/ml

Type: Primary
Antigen: BAX
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR