Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-BAK Rabbit Polyclonal Antibody

Supplier: BioVision

3031-100
10003-252EA 252.35 USD InStock
10003-252
Anti-BAK Rabbit Polyclonal Antibody
Antibodies
Bak is a homolog of apoptosis inhibitor Bcl-2. Bak is a member of the Bcl-2 family of proteins and contains Bcl-2 homology region (BH-) 1, 2 and 3. Unlike Bcl-2, Bak promotes cell death and counteracts the anti-apoptotic effect of Bcl-2. The immunoaffinity purified antibody detects a 30 kDa band, corresponding to the expected molecular weight of Bak on SDS-PAGE immunoblots.

Recommended Dilutions: Western Blot: 1-4 µg/ml; Immunoprecipitation: 5-20 µg/ml).

Type: Primary
Antigen: BAK
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty