Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-BAD Rabbit Polyclonal Antibody

Supplier: BioVision

3030-100
10003-250EA 272.95 USD InStock
10003-250
Anti-BAD Rabbit Polyclonal Antibody
Antibodies
The Bcl-2 (B-cell leukemia/lymphoma 2) family proteins function to either inhibit (Bcl-2 and Bcl-XL) or promote (Bax and Bad) apoptosis. Similar to other family member, Bad also exhibits sequence homology to Bcl-2. Bad can selectively dimerize with Bcl-xL and Bcl-2, but not with Bax, Bcl-xs, Mcl-1, or itself. In mammalian cells, Bad binds more strongly to Bcl-xL than Bcl-2. This may explain why Bad reverse the death repressor activity of Bcl-xL, but not that of Bcl-2.

Recommended Dilutions: Western Blot: 1-2 µg/ml; Immunoprecipitation: 4-8 µg/ml; and Immunohistochemistry: 20-40 µg/ml

Type: Primary
Antigen: BAD
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Monkey

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty