Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CASP5 Rabbit Polyclonal Antibody

Supplier: BioVision

3029-100
10003-246EA 252.35 USD InStock
10003-246
Anti-CASP5 Rabbit Polyclonal Antibody
Antibodies
Caspase family of cysteine proteases has been shown to play a key role in apoptosis. Caspase-5 is also known as Ich-3 and ICErel-III and can proteolytically processed by granzyme B. Over expression of caspase-5 induces apoptosis in Rat-1 and HeLa cells, which can be inhibited by CrmA and Bcl-2.

Recommended Dilutions: Western Blot: 0.5-4ag/ml

Type: Primary
Antigen: CASP5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human, Mouse, Rat, Monkey, Hamster, Bovine

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty