Anti-CASP2 Rabbit Polyclonal Antibody

Supplier: BioVision
3027-100
10003-238EA 297.72 USD
10003-238
Anti-CASP2 Rabbit Polyclonal Antibody
Caspases play a key role in apoptosis. Similar as other caspases, caspase-2 is also synthesized as an inactive pro-enzyme that is processed in cells undergoing apoptosis. Caspase-2 mRNA is alternatively spliced. The larger mRNA species encoding a product of 435 amino acids is known as ICH-1L. The smaller mRNA species encoding a protein of 312 amino acids is named ICH-1S. Over expression of ICH-1L induces apoptosis, while over expression of ICH-1S suppresses Rat-1 cell death induced by serum deprivation.

Recommended Dilutions: Western Blot: 1-2 µg/ml; and Immunohistochemistry: 5 µg/ml

Type: Primary
Antigen: CASP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR