Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CYCS Mouse Monoclonal Antibody [clone: 7H8.2C12]

Supplier: BioVision

3026-100
10003-236EA 375.95 USD InStock
10003-236
Anti-CYCS Mouse Monoclonal Antibody [clone: 7H8.2C12]
Antibodies
Cytochrome c (m.w. 12,500) is an electron transport protein from mitochondria. It is released from mitochondria to cytoplasm during the early stages of apoptosis, prior to caspase activation, DNA fragmentation, and loss of membrane potential. The cytoplases.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: CYCS
Clonality: Monoclonal
Clone: 7H8.2C12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty